0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
Elektrokemiske instrumenter
pH-metre
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
Nitrat-måleelektrode, VARiON NO3
706-0507
:
WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
706-0507
:
WTW ONLINE
10+
Accessories for on-line multi-parameter systems, IQ sensor net
Nitrat-måleelektrode, VARiON NO3
4.490,00 kr
1 * 1 Items
-
+
Beskrivelse:
Nitrat-måleelektrode, VARiON NO3
Seq:
18
Kaliumhexacyanoferrat (III) ≥99.0%, AnalaR NORMAPUR® analytical reagent
#: 26810.298
: VWR Chemicals