Volumetric flasks, class A, white markings
150 ml, 14/23, 0,150 ± ml
Katalog # WITG3.670.150
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3, amber, with glass stopper.
- White markings
- Conformity certified (DE-M marking)
- Inclusive batch certificate
Certifications: DIN EN ISO 1042
Specifikationer
- NS:14/23
- Kapacitet:150 ml
- Seq:07
- Tolerance:0,150 ± ml