Orders LinkContactUsLinkComponent
Robotic OMIX™ tips for Tomtec Quadra
Robotic OMIX™ tips for Tomtec Quadra
OMIX MP1, -, Tomtec Quadra, 5 racks, 96 tips each
Katalog # VARIA57311
Leverandør:   VARIAN
Robotic OMIX™ tips for Tomtec Quadra
Katalog # VARIA57311
Leverandør:  VARIAN
Robotic OMIX™ tips for Tomtec Quadra
OMIX MP1, -, Tomtec Quadra, 5 racks, 96 tips each
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These Tomtec compatible tips contain a slice of monolithic SPE material.This allows for vacuum-free processing and walk-away automation. With hands-free SPE the process becomes much more streamlined and reproducible.
  • Fast, uniform flow maximises productivity and reproducibility
  • Small monolithic tip delivers low elution volumes, increasing sensitivity and reducing solvent usage
  • Vacuum-free processing improves reproducibility and shortens processing times
Specifikationer
  • Pakket:
    5 racks, 96 tips each
  • Spidstype:
    OMIX MP1
  • Til brug med:
    Tomtec Quadra
  • Filter:
    -