Robotic OMIX™ tips for Tomtec Quadra
OMIX MP1, -, Tomtec Quadra, 5 racks, 96 tips each
Katalog # VARIA57311
Leverandør: VARIAN
Gå til:
Product Details & Documents
These Tomtec compatible tips contain a slice of monolithic SPE material.This allows for vacuum-free processing and walk-away automation. With hands-free SPE the process becomes much more streamlined and reproducible.
- Fast, uniform flow maximises productivity and reproducibility
- Small monolithic tip delivers low elution volumes, increasing sensitivity and reducing solvent usage
- Vacuum-free processing improves reproducibility and shortens processing times
Specifikationer
- Pakket:5 racks, 96 tips each
- Spidstype:OMIX MP1
- Til brug med:Tomtec Quadra
- Filter:-