Flame atomisation systems
Burner cleaning and alignment card assembly
Katalog # VARI9910053900
Leverandør: VARIAN
Gå til:
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
- Contoured design for outstanding resistance to blockage even with the most difficult examples
- Corrosion-resistant components
- For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specifikationer
- Beskrivelse:Burner cleaning and alignment card assembly
- Seq:303
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"VARI6910009700Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Glass impact beads
Produktinformation
- Katalog #TilgængelighedPrice
- Aqueous spray chamber o-ring
Produktinformation
- Katalog #TilgængelighedPrice
- Organics o-ring
Produktinformation
- Katalog #TilgængelighedPrice
- Fluorinated mixing paddles
Produktinformation
- Katalog #TilgængelighedPrice
- Integrated liquid trap (fluorinated float and magnet)Integrated liquid trap (fluorinated float and magnet)VARI9910105800Log ind for tilgængelighed1 * 1 Items - 4.190,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Nebuliser capillary kit
Produktinformation
- Katalog #TilgængelighedPrice
- Integrated nebuliser block
Produktinformation
- Katalog #TilgængelighedPrice
- Flame atomisation system kit