Orders LinkContactUsLinkComponent
Flame atomisation systems
Flame atomisation systems
High vacuum preset barrel nebuliser
Catalog # VARI9910043700
Supplier:   VARIAN
Flame atomisation systems
Catalog # VARI9910043700
Supplier:  VARIAN
Flame atomisation systems
Flame atomisation systems
High vacuum preset barrel nebuliser
Price
17.930,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specifications
  • Beskrivelse:
    High vacuum preset barrel nebuliser
  • Seq:
    204
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI6910009700
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    Log in for availability
    1 * 1 Items
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910025700
    Glass impact beads
    Log in for availability
    1 * 5 Items - 2.010,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910093400
    Aqueous spray chamber o-ring
    Log in for availability
    1 * 1 Items - 846,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910093500
    Organics o-ring
    Log in for availability
    1 * 1 Items - 11.490,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910093600
    Fluorinated mixing paddles
    Log in for availability
    1 * 1 Items - 2.480,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910105800
    Integrated liquid trap (fluorinated float and magnet)
    Log in for availability
    1 * 1 Items - 4.190,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910093000
    Nebuliser capillary kit
    Log in for availability
    1 * 1 Items - 7.550,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI9910093100
    Integrated nebuliser block
    Log in for availability
    1 * 1 Items - 13.360,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • VARI190034100
    Flame atomisation system kit
    Log in for availability
    1 * 1 Items - 28.370,00 kr