Fluorescence cuvettes
Sort væg, Semi-micro, 450 µl, 5 mm, UV-kvartsglas
VARI6610019800
: VARIAN
Fluorescence cuvettes designed for UV/visible analysis in routine applications of molecular spectroscopy.
- Longer pathlength cells for extra sensitivity in low concentration samples
Suitable for low concentration or highly corrosive samples.
Longer pathlength (20 - 50 mm) cells are suitable for extra sensitivity in low concentration samples.
Ordering information: 10 mm pathlength models are available in matched pairs.
- Længde på lysvej:5 mm
- Model:Semi-micro
- Volumen:450 µl
- Vinduesmateriale:UV-kvartsglas
- Beskrivelse:Sort væg