Fluorescence cuvettes
Semimikro-celle, sort væg, Semi-micro, 90 µl, 1 mm, UV-kvartsglas
Katalog # VARI6610019600
Leverandør: VARIAN
Gå til:
Product Details & Documents
Fluorescence cuvettes designed for UV/visible analysis in routine applications of molecular spectroscopy.
- Longer pathlength cells for extra sensitivity in low concentration samples
Suitable for low concentration or highly corrosive samples.
Longer pathlength (20 - 50 mm) cells are suitable for extra sensitivity in low concentration samples.
Ordering information: 10 mm pathlength models are available in matched pairs.
Specifikationer
- Længde på lysvej:1 mm
- Model:Semi-micro
- Volumen:90 µl
- Vinduesmateriale:UV-kvartsglas
- Beskrivelse:Semimikro-celle, sort væg
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Cellefod
Produktinformation
- Katalog #TilgængelighedPrice
- Magnetomrørerstave, 7×2 mm
Produktinformation
- Katalog #TilgængelighedPrice
- Magnetomrørerstav, PTFE