Fluorescence cuvettes
Standardcelle, åben top, 17,5 ml, 50 mm, Glas
VARI6610016400
: VARIAN
Fluorescence cuvettes designed for UV/visible analysis in routine applications of molecular spectroscopy.
- Longer pathlength cells for extra sensitivity in low concentration samples
Suitable for low concentration or highly corrosive samples.
Longer pathlength (20 - 50 mm) cells are suitable for extra sensitivity in low concentration samples.
Ordering information: 10 mm pathlength models are available in matched pairs.
- Længde på lysvej:50 mm
- Volumen:17,5 ml
- Vinduesmateriale:Glas
- Beskrivelse:Standardcelle, åben top