n-Hexan, EMSURE® ACS for analysis, Supelco®
Plastic bottle, 2,5 L
Leverandør: Merck
n-Hexan, EMSURE® ACS for analysis, Supelco®
Katalog # 1.04367.2511
Leverandør: Merck
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :M-Clarity™ quality level = MQ300
- EINECS:203-777-6
Documents
Certifikater
Søg efter partinummerMERC1.04367.2500_TDS_EN ProductPage_104367 ProductPage_104367
Produktspecifikation
Merck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure
Produktlitteratur
msds_104367
SDS
Specifikationer
Specifikationstestresultater
- Purity (GC):≥ 99.0 %
- Identity (IR):conforms
- Color:≤ 10 Hazen
- Alkalinity:≤ 0.0002 meq/g
- Acidity:≤ 0.0002 meq/g
- Density (d 20 °C/ 4 °C):0.659 - 0.662
- Thiophene:conforms
- Aromatics (as benzene):≤ 0.01 %
- Sulfur compounds (as S):≤ 0.005 %
- Matter discolored by H₂SO₄:≤ 10 Hazen
- Al (Aluminium):≤ 0.00005 %
- B (Boron):≤ 0.000002 %
- Ba (Barium):≤ 0.00001 %
- Ca (Calcium):≤ 0.00005 %
- Cd (Cadmium):≤ 0.000005 %
- Co (Cobalt):≤ 0.000002 %
- Cr (Chromium):≤ 0.000002 %
- Cu (Copper):≤ 0.000002 %
- Fe (Iron):≤ 0.00001 %
- Mg (Magnesium):≤ 0.00001 %
- Mn (Manganese):≤ 0.000002 %
- Ni (Nickel):≤ 0.000002 %
- Pb (Lead):≤ 0.00001 %
- Sn (Tin):≤ 0.00001 %
- Zn (Zinc):≤ 0.00001 %
- Evaporation residue:≤ 0.001 %
- Water:≤ 0.005 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



