Orders LinkContactUsLinkComponent
Ginger, Reference Standards
Ginger, Reference Standards
Ginger Constituent Mixture, USP reference standard, Vial, 0,6 mg
Katalog # 1291446.
Leverandør:   USP
Ginger, Reference Standards
Katalog # 1291446.
Leverandør:  USP
CAS nummer:  
Ginger, Reference Standards
Ginger Constituent Mixture, USP reference standard, Vial, 0,6 mg
CAS nummer:  
Price
4.410,00 kr
Log ind for tilgængelighed
1 * 0,6 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Ginger Constituent Mixture: A freeze-dried mixture of 6-Gingerol and 6-Shogaol
Certifications: US Pharmacopea Reference Standard
Specifikationer
Specifikationer
  • Pakning:
    Vial
  • Pakn:
    0,6 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Ginger Constituent Mixture