Glass - trace elements, NIST standards
NIST-616 glass - trace elements, 1
Catalog # PROONIST-616
Supplier: LGC Standards PROMOCHEM
Glass - trace elements, NIST standards
Catalog # PROONIST-616
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifications
Specifications
- Pakn:1
- Beskrivelse:NIST-616 glass - trace elements