Glass - trace elements, NIST standards
NIST-613 glass - trace elements, 1
Katalog # PROONIST-613
Leverandør: LGC Standards PROMOCHEM
Glass - trace elements, NIST standards
Katalog # PROONIST-613
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifikationer
Specifikationer
- Pakn:1
- Beskrivelse:NIST-613 glass - trace elements