Glass - trace elements, NIST standards
NIST- 611, glass - trace elements, thickness 1 mm, 1
Katalog # PROONIST-611
Leverandør: LGC Standards PROMOCHEM
Glass - trace elements, NIST standards
Katalog # PROONIST-611
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifikationer
Specifikationer
- Pakn:1
- Beskrivelse:NIST- 611, glass - trace elements, thickness 1 mm