Orders LinkContactUsLinkComponent
Glass - trace elements, NIST standards
Glass - trace elements, NIST standards
NIST- 610, glass - trace elements, thickness 3 mm, 1
Katalog # PROONIST-610
Leverandør:   LGC Standards PROMOCHEM
Glass - trace elements, NIST standards
Katalog # PROONIST-610
CAS nummer:  
Glass - trace elements, NIST standards
NIST- 610, glass - trace elements, thickness 3 mm, 1
CAS nummer:  
Price
12.200,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    NIST- 610, glass - trace elements, thickness 3 mm