Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Hydroquinon 99.5%

Glass bottle for solids, 5 g
Catalog # ACRO219930050
Irritant
Health
Corrosive
Environment
Hydroquinon 99.5%
Catalog # ACRO219930050
CAS Number:  123-31-9
Hydroquinon 99.5%
Hydroquinon 99.5%

Glass bottle for solids, 5 g

CAS Number:  123-31-9
Price
256,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1,4-Dihydroxybenzen, Benzen-1,4-diol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,04833
  • Boiling Pt:285…287 °C (760 mmHg)
  • Melting Pt:170…174 °C
  • Density:1,332 g/cm³ (20 °C)
  • Flash Pt:165 °C
  • Storage Temperature:Stue temperatur
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    171 to 174 °C
  • Water:
    ≤0.6 % (K.F.)
  • Sulfated ash:
    ≤0.05 %
  • HPLC:
    ≥99.4 %

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifications