2-Propanol 99.5% for analysis
Metal drum, 25 L
Leverandør: Thermo Fisher Scientific
2-Propanol 99.5% for analysis
Katalog # ACRO184130250
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Kogende Pt:81…83 °C (760 mmHg)
- Smeltende Pt:–89,5 °C
- Tæthed:0,785 g/cm³
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1219
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.1 % (Coulometric)
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0001 meq/g corresp. 0.0007% C3H6O2
- Substance darkened by H2SO4:≤10 APHA
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Sodium (Na):≤1 ppm
- Sulfur (S):≤0.5 ppm
- Zinc (Zn):≤0.1 ppm
Specifikationer
- Pakning:Metal drum
- Pakn:25 L
- Cat. No.:ACRO184130250
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



