Natriumhydroxid, micropearls for analysis
Plastic bottle, 250 g
Leverandør: Thermo Fisher Scientific
Natriumhydroxid, micropearls for analysis
Katalog # ACRO383042500
Leverandør: Thermo Fisher Scientific
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kogende Pt:1390 °C (1013 hPa)
- Smeltende Pt:318 °C
- Tæthed:2,13 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- NÅ:01-2119457892-27
Documents
ACRO38304
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Pearls
- Titration with HCl:≥99.0 %
- Sodium carbonate (Na2CO3):≤0.4 %
- Sodium sulfate (Na2SO4):≤80 ppm
- Sodium chloride (NaCl):≤200 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤0.5 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



