n-Hexan, extra dry 96+%, AcroSeal®
Glass bottle, 100 mL
Leverandør: Thermo Fisher Scientific
n-Hexan, extra dry 96+%, AcroSeal®
Katalog # 326921000.
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (760 mmHg)
- Smeltende Pt:–95 °C
- Tæthed:0,659 g/cm³
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Water:≤0.002 %
- GC:≥96.0 %
- Residue after evaporation:≤0.0005 %
- Filt.over 0.2 micron PFTE:Filtered
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



