2-Propanol 99.5%, gradient grade for HPLC
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
2-Propanol 99.5%, gradient grade for HPLC
Katalog # ACRO383920025
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Kogende Pt:81…83 °C (760 mmHg)
- Smeltende Pt:–89,5 °C
- Tæthed:0,785 g/cm³
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-661-7
- FN:1219
- ADR:3,II
- NÅ:01-2119457558-25
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Water:≤0.1 %
- GC:≥99.4 %
- UV:at 260 nm A: ≤0.005
- Residue after evaporation:≤2 ppm
- Acidity/alkalinity:≤0.0001 meq/g
- Gradient analysis::
- Absorbance largest eluted peak:≤0.002 A.U.
- Filtered to 0.2 micrometer:Filtered
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:ACRO383920025
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



