n-Pentan for analysis
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
n-Pentan for analysis
Katalog # ACRO383900010
Leverandør: Thermo Fisher Scientific
CAS nummer: 109-66-0
Gå til:
Product Details & Documents
Synonymer: Pentan
- CAS nummer:109-66-0
- Formel:H₃C(CH₂)₃CH₃
- MDL nummer:MFCD00009498
- Molecular Weight:72.15 g/mol
- Merck Index:13,07193
- Kogende Pt:36 °C (760 mmHg)
- Smeltende Pt:–130 °C
- Tæthed:0,626 g/cm³
- Flash Pt:–49 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-692-4
- FN:1265
- ADR:3,II
- NÅ:01-2119459286-30
Documents
ACRO38390
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.01 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Acidity/alkalinity:≤0.0001 meq/g
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.2 ppm
- Lead (Pb):≤0.05 ppm
- Magnesium (Mg):≤0.1 ppm
- Potassium (K):≤0.2 ppm
- Sodium (Na):≤1 ppm
- Zinc (Zn):≤0.1 ppm
- Total phosphorus (P):≤0.5 ppm
- Total silicon (Si):≤0.02 ppm
- Total sulfur (S):≤5 ppm
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L



