n-Hexan contains appr. 95% n-Hexane for analysis
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
n-Hexan contains appr. 95% n-Hexane for analysis
Katalog # ACRO383800025
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (760 mmHg)
- Smeltende Pt:–95 °C
- Tæthed:0,659 g/cm³
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Documents
ACRO38380
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.02 % (K.F.)
- GC:≥95.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Acidity/alkalinity:≤0.00008 meq/g
- Substance darkened by H2SO4:passes test
- Calcium (Ca):≤0.2 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Hydrocarbons:≤0.1 % Aromatic hydrocarbons
- Total phosphorus (P):≤0.1 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤10 ppm
- Bromine number:≤0.5
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



