n-Hexan contains appr. 95% n-Hexane for analysis
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
n-Hexan contains appr. 95% n-Hexane for analysis
Katalog # ACRO383800010
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (760 mmHg)
- Smeltende Pt:–95 °C
- Tæthed:0,659 g/cm³
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Documents
ACRO38380
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.02 % (K.F.)
- GC:≥95.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Acidity/alkalinity:≤0.00008 meq/g
- Substance darkened by H2SO4:passes test
- Calcium (Ca):≤0.2 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Hydrocarbons:≤0.1 % Aromatic hydrocarbons
- Total phosphorus (P):≤0.1 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤10 ppm
- Bromine number:≤0.5
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



