Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dichlormethan 99+% stabilised, extra pure

Dichlormethan 99+% stabilised, extra pure

Metal drum, 25 L
Catalog # ACRO383780250
Irritant
Health
Dichlormethan 99+% stabilised, extra pure
Catalog # ACRO383780250
CAS Number:  75-09-2
Dichlormethan 99+% stabilised, extra pure
Dichlormethan 99+% stabilised, extra pure

Metal drum, 25 L

CAS Number:  75-09-2
Price
3.960,00 kr
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Methylenchlorid
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39…40 °C (760 mmHg)
  • Melting Pt:–97 °C
  • Density:1,325 g/cm³
  • Storage Temperature:Stue temperatur
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :stabiliseret med amylen
  • UN:1593

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (K.F.)
  • GC:
    ≥99.0 %
  • Stabilizer:
    Approx. 100 ppm Amylene
  • Residue after evaporation:
    ≤0.05 %

Specifications

  • Pakning:
    Metal drum
  • Pakn:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.