Agar, powder, pure
Glass bottle for solids, 1 kg
Leverandør: Thermo Fisher Scientific
Agar, powder, pure
Katalog # ACRO400400010
Leverandør: Thermo Fisher Scientific
CAS nummer: 9002-18-0
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Stue temperatur
- EINECS:232-658-1
Documents
ACRO40040
SDS
Specifications
Specification Test Results
- Appearance:White to cream to light yellow to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤20.0 % (1 g, 105°C, 5 h)
- Heavy metals:≤20 ppm
- Gel strength:500 to 1000 g/cm² (1.5 % at 20°C)
- Ash:≤6 %
- Microbial contamination:≤1000 /g (Total plate count)
- pH:5.7 to 7.5 (1.5 % in water)
Specifications
- Pakning:Glass bottle for solids
- Pakn:1 kg



