Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Dichlormethan 99.5% stabilised for analysis

Glass bottle, 2,5 L
  ACRO390700025
 
 
Dichlormethan 99.5% stabilised for analysis
  ACRO390700025
 :  75-09-2
Dichlormethan 99.5% stabilised for analysis
Dichlormethan 99.5% stabilised for analysis

Glass bottle, 2,5 L

 :  75-09-2
 
823,00 kr
   
1 * 2,5 L
 
 :  Methylenchlorid
  •  :75-09-2
  •  :CH₂Cl₂
  •  :MFCD00000881
  •  :84.93 g/mol
  •  :12,06140
  •  :39…40 °C (760 mmHg)
  •  :–97 °C
  •  :1,325 g/cm³
  •  :Stue temperatur
  •  :200-838-9
  •  :6.1,III
  •  :stabiliseret med ethanol
  •  :1593

 

 

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 % (K.F.)
  • GC:
    ≥99.4 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.0005 meq/g
  • Free chlorine:
    ≤0.00003 % (as Cl)
  • Aluminium (Al):
    ≤0.5 ppm
  • Barium (Ba):
    ≤0.1 ppm
  • Cadmium (Cd):
    ≤0.05 ppm
  • Calcium (Ca):
    ≤0.5 ppm
  • Chromium (Cr):
    ≤0.02 ppm
  • Cobalt (Co):
    ≤0.02 ppm
  • Copper (Cu):
    ≤0.02 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Manganese (Mn):
    ≤0.02 ppm
  • Nickel (Ni):
    ≤0.02 ppm
  • Tin (Sn):
    ≤0.1 ppm
  • Zinc (Zn):
    ≤0.1 ppm
  • Aldehyde (HCHO):
    ≤0.0005 %
  • Ethanol:
    ≤0.3 % (GC)

 

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L