Dichlormethan 99.5% stabilised for analysis
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Dichlormethan 99.5% stabilised for analysis
Katalog # ACRO390700010
Leverandør: Thermo Fisher Scientific
CAS nummer: 75-09-2
Gå til:
Product Details & Documents
Synonymer: Methylenchlorid
- CAS nummer:75-09-2
- Formel:CH₂Cl₂
- MDL nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Kogende Pt:39…40 °C (760 mmHg)
- Smeltende Pt:–97 °C
- Tæthed:1,325 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-838-9
- ADR:6.1,III
- :stabiliseret med ethanol
- FN:1593
Documents
ACRO39070
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.02 % (K.F.)
- GC:≥99.4 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.0005 meq/g
- Free chlorine:≤0.00003 % (as Cl)
- Aluminium (Al):≤0.5 ppm
- Barium (Ba):≤0.1 ppm
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Tin (Sn):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Aldehyde (HCHO):≤0.0005 %
- Ethanol:≤0.3 % (GC)
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



