Methylethylketon 99.7% for HPLC
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Methylethylketon 99.7% for HPLC
Katalog # ACRO389570010
Leverandør: Thermo Fisher Scientific
CAS nummer: 78-93-3
Gå til:
Product Details & Documents
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:13,06097
- Kogende Pt:80 °C (760 mmHg)
- Smeltende Pt:–87 °C
- Tæthed:0,806 g/cm³
- Flash Pt:–7 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Documents
ACRO38957
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.6 %
- UV:at 330 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0003 meq/g
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
- Cat. No.:ACRO389570010
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



