Methylethylketon ACS
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
Methylethylketon ACS
Katalog # 389430025.
Leverandør: Thermo Fisher Scientific
CAS nummer: 78-93-3
Gå til:
Product Details & Documents
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:13,06097
- Kogende Pt:80 °C (760 mmHg)
- Smeltende Pt:–87 °C
- Tæthed:0,806 g/cm³
- Flash Pt:–7 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Documents
ACRO38943
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.20 %
- GC:≥99.0 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.0025 %
- Titrable acid:≤0.0005 meq/g
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:389430025.
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



