Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Jod 99+%, pure

Jod 99+%, pure

Glass bottle, 250 g
Katalog # ACRO387052500
Irriterende
Sundhed
Miljø
Leverandør:   Thermo Fisher Scientific
Jod 99+%, pure
Katalog # ACRO387052500
CAS nummer:  7553-56-2
Jod 99+%, pure
Jod 99+%, pure

Glass bottle, 250 g

CAS nummer:  7553-56-2
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
  • CAS nummer:7553-56-2
  • Formel:I₂
  • MDL nummer:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:13,05036
  • Kogende Pt:185 °C (760 mmHg)
  • Smeltende Pt:113 °C
  • Tæthed:4,9 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-442-4
  • FN:3495
  • ADR:8,III

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark grey Metallic spheres
  • Titration Iodometric:
    ≥99.0 %
  • Insoluble matter:
    Pass

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.