Jod 99+%, pure
Glass bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:7553-56-2
- Formel:I₂
- MDL nummer:MFCD00011355
- Molecular Weight:253.81 g/mol
- Merck Index:13,05036
- Kogende Pt:185 °C (760 mmHg)
- Smeltende Pt:113 °C
- Tæthed:4,9 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-442-4
- FN:3495
- ADR:8,III
Documents
ACRO38705
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Dark grey Metallic spheres
- Titration Iodometric:≥99.0 %
- Insoluble matter:Pass
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



