Triethylamin 99%, pure
Glass bottle, 100 mL
Leverandør: Thermo Fisher Scientific
Triethylamin 99%, pure
Katalog # ACRO157911000
Leverandør: Thermo Fisher Scientific
CAS nummer: 121-44-8
Gå til:
Product Details & Documents
Synonymer: (Diethylamino)ethane, TEA
- CAS nummer:121-44-8
- Formel:(C₂H₅)₃N
- MDL nummer:MFCD00009051
- Molecular Weight:101.19 g/mol
- Merck Index:13,09740
- Kogende Pt:90 °C (760 mmHg)
- Smeltende Pt:–115 °C
- Tæthed:0,728 g/cm³
- Flash Pt:–11 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-469-4
- FN:1296
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3990 to 1.4010 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL
- Cat. No.:ACRO157911000



