Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Triethylamin 99%, pure

Glass bottle, 5 mL
Katalog # ACRO157910050
Giftig
Ætsende
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Triethylamin 99%, pure
Katalog # ACRO157910050
CAS nummer:  121-44-8
Triethylamin 99%, pure
Triethylamin 99%, pure

Glass bottle, 5 mL

CAS nummer:  121-44-8
Price
292,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
Synonymer:  (Diethylamino)ethane, TEA
  • CAS nummer:121-44-8
  • Formel:(C₂H₅)₃N
  • MDL nummer:MFCD00009051
  • Molecular Weight:101.19 g/mol
  • Merck Index:13,09740
  • Kogende Pt:90 °C (760 mmHg)
  • Smeltende Pt:–115 °C
  • Tæthed:0,728 g/cm³
  • Flash Pt:–11 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-469-4
  • FN:1296
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3990 to 1.4010 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 mL

Specifikationer