Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Propanol 99.5+%

2-Propanol 99.5+%

Plastic drum, 10 L
Katalog # ACRO149320100
Irriterende
Brandfarlig
Leverandør:   Thermo Fisher Scientific
2-Propanol 99.5+%
Katalog # ACRO149320100
CAS nummer:  67-63-0
2-Propanol 99.5+%
2-Propanol 99.5+%

Plastic drum, 10 L

CAS nummer:  67-63-0
Price
2.440,00 kr
Log ind for tilgængelighed
1 * 10 L
Product Details & Documents
Synonymer:  Isopropylalkohol
  • CAS nummer:67-63-0
  • Formel:(CH₃)₂CHOH
  • MDL nummer:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Kogende Pt:82 °C (1013 hPa)
  • Smeltende Pt:–89,5 °C
  • Tæthed:0,785 g/cm³
  • Flash Pt:12 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-661-7
  • FN:1219
  • ADR:3,II
  • NÅ:01-2119457558-25

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥99.50 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    10 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.