Diethylether 99+% stabilised, pure
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
Diethylether 99+% stabilised, pure
Katalog # ACRO123990025
Leverandør: Thermo Fisher Scientific
CAS nummer: 60-29-7
Gå til:
Product Details & Documents
Synonymer: Ether
- CAS nummer:60-29-7
- Formel:(CH₃CH₂)₂O
- MDL nummer:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Kogende Pt:34,6 °C (760 mmHg)
- Smeltende Pt:–116 °C
- Tæthed:0,714 g/cm³
- Flash Pt:–45 °C
- FN:1155
- ADR:3,I
- :stabiliseret med BHT/Ionol
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-467-2
- NÅ:01-2119535785-29
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3510 to 1.3540 (20°C, 589 nm)
- Water:≤0.2 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 10 ppm BHT
- Residue after evaporation:≤0.002 %
- Peroxides:≤0.001 %
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



