Orders LinkContactUsLinkComponent
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffer I, includes urea and Nonidet® P-40, 50 mL
Katalog # 786-220
Leverandør:   G-Biosciences
Extraction buffers, FOCUS™
Katalog # 786-220
Leverandør:  G-Biosciences
CAS nummer:  
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffer I, includes urea and Nonidet® P-40, 50 mL
CAS nummer:  
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 50 mL
Product Details & Documents
FOCUS™ Extraction Buffers are experimentally optimised concentrations of critical agents, buffering and stabilising agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulphobetaines (SB). They are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.
Specifikationer
Specifikationer
  • Pakn:
    50 mL
  • Beskrivelse:
    FOCUS™ Extraction Buffer I, includes urea and Nonidet® P-40
Specifikationer