Orders LinkContactUsLinkComponent
Single tissue blots, multiple species
Single tissue blots, multiple species
Human, mouse and rat testis single tissue, multiple species blot, 1
Catalog # TB32
Supplier:   G-Biosciences
Single tissue blots, multiple species
Catalog # TB32
Supplier:  G-Biosciences
CAS Number:  
Single tissue blots, multiple species
Single tissue blots, multiple species
Human, mouse and rat testis single tissue, multiple species blot, 1
CAS Number:  
Price
4.640,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Single tissue and multiple species blot contains four protein lanes with an identical GenLysate™ tissue lysate from human, mouse and rat species, and a protein marker lane.
  • Ready to screen
  • Single tissue multiple species (human, mouse, rat) blot
The premade blots are prepared with the GenLysate™ and then 50μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
Specifications
Specifications
  • Pakn:
    1
  • Beskrivelse:
    Human, mouse and rat testis single tissue, multiple species blot