Orders LinkContactUsLinkComponent
Protein purification kit, STRATEGY™
Protein purification kit, STRATEGY™
Anionic resin columns, 6
Katalog # 786-184A
Leverandør:   G-Biosciences
Protein purification kit, STRATEGY™
Katalog # 786-184A
Leverandør:  G-Biosciences
CAS nummer:  
Protein purification kit, STRATEGY™
Protein purification kit, STRATEGY™
Anionic resin columns, 6
CAS nummer:  
Price
1.270,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
STRATEGY™ is designed to allow researchers to rapidly screen a number of purification techniques in a small scale format to identify the system to isolate their protein of interest. The self-contained kit allows researchers to save time and money as separate buffers, resins and columns are not needed.
The kit guides the researcher step-by-step through various protein purification techniques like pH fractionation, salt fractionation, hydrophobic chromatography, anionic chromatography and cationic chromatography.
Specifikationer
Specifikationer
  • Pakn:
    6
  • Beskrivelse:
    Anionic resin columns