Protein purification kit, STRATEGY™
Anionic resin columns, 6
Katalog # 786-184A
Leverandør: G-Biosciences
Gå til:
Product Details & Documents
STRATEGY™ is designed to allow researchers to rapidly screen a number of purification techniques in a small scale format to identify the system to isolate their protein of interest. The self-contained kit allows researchers to save time and money as separate buffers, resins and columns are not needed.
The kit guides the researcher step-by-step through various protein purification techniques like pH fractionation, salt fractionation, hydrophobic chromatography, anionic chromatography and cationic chromatography.
Documents
Specifikationer
Specifikationer
- Pakn:6
- Beskrivelse:Anionic resin columns