Orders LinkContactUsLinkComponent
Inclusion bodies solubilisation buffer
Inclusion bodies solubilisation buffer
IBS-HP™ buffer, 1 KIT
Catalog # 786-183HP
Supplier:   G-Biosciences
Inclusion bodies solubilisation buffer
Catalog # 786-183HP
Supplier:  G-Biosciences
CAS Number:  
Inclusion bodies solubilisation buffer
Inclusion bodies solubilisation buffer
IBS-HP™ buffer, 1 KIT
CAS Number:  
Price
2.140,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
The IBS™ buffer is specifically developed for solubilisation of inclusion bodies.
IBS-HP™ Buffer is for the solubilisation of inclusion bodies containing highly hydrophobic proteins.
Specifications
Specifications
  • Pakn:
    1 KIT
  • Beskrivelse:
    IBS-HP™ buffer