Orders LinkContactUsLinkComponent
Inclusion bodies solubilisation buffer
Inclusion bodies solubilisation buffer
IBS™ buffer, 100 mL
Katalog # 786-183
Leverandør:   G-Biosciences
Inclusion bodies solubilisation buffer
Katalog # 786-183
Leverandør:  G-Biosciences
CAS nummer:  
Inclusion bodies solubilisation buffer
Inclusion bodies solubilisation buffer
IBS™ buffer, 100 mL
CAS nummer:  
Price
2.140,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
The IBS™ buffer is specifically developed for solubilisation of inclusion bodies.
IBS-HP™ Buffer is for the solubilisation of inclusion bodies containing highly hydrophobic proteins.
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Beskrivelse:
    IBS™ buffer
Specifikationer