Orders LinkContactUsLinkComponent
G-Biosciences TAE Running Buffer (50X)
G-Biosciences TAE Running Buffer (50X)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Plastic bottle, 1 L
Katalog # R023
Leverandør:   G-Biosciences
G-Biosciences TAE Running Buffer (50X)
Katalog # R023
Leverandør:  G-Biosciences
CAS nummer:  
G-Biosciences TAE Running Buffer (50X)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Plastic bottle, 1 L
CAS nummer:  
Price
kr721.00
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
The primary use of TAE Running Buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
  • 50X Concentrated solution: 2 M Tris-Acetate, 0,05 M EDTA (pH 8,3)
TAE has a lower buffer capacity than TBE Running Buffer and can easily become exhausted, however linear, double stranded DNA runs faster on TAE.
Specifikationer
Specifikationer
  • Pakning:
    Plastic bottle
  • Pakn:
    1 L
  • Beskrivelse:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)