G-Biosciences TAE Running Buffer (50X)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Plastic bottle, 1 L
Katalog # R023
Leverandør: G-Biosciences
Gå til:
Product Details & Documents
The primary use of TAE Running Buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
50X Concentrated solution: 2 M Tris-Acetate, 0,05 M EDTA (pH 8,3)
TAE has a lower buffer capacity than TBE Running Buffer and can easily become exhausted, however linear, double stranded DNA runs faster on TAE.
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 L
- Beskrivelse:TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)