Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Vand, DEPC treated

Vand, DEPC treated

Plastic bottle, 500 mL
Katalog # 786-118
Leverandør:   G-Biosciences
Vand, DEPC treated
Katalog # 786-118
Leverandør:  G-Biosciences
CAS nummer:  7732-18-5
Vand, DEPC treated
Vand, DEPC treated

Plastic bottle, 500 mL

CAS nummer:  7732-18-5
Price
395,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Ultra pure DEPC-Treated water is suitable for use with sensitive RNA research. It is prepared by incubating with 0.1% DEPC (diethylpyrocarbonate), an RNase inhibitor, and then autoclaved to remove the DEPC, to prevent downstream interference, and sterilized.

Synonymer:  Hydrogen oxide
  • CAS nummer:7732-18-5
  • Formel:H₂O
  • MDL nummer:MFCD00011332
  • Molecular Weight:18.02 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL

Specifikationer