Vand, DEPC treated
Plastic bottle, 125 mL
786-117
Ultra pure DEPC-Treated water is suitable for use with sensitive RNA research. It is prepared by incubating with 0.1% DEPC (diethylpyrocarbonate), an RNase inhibitor, and then autoclaved to remove the DEPC, to prevent downstream interference, and sterilized.
: Hydrogen oxide
- :7732-18-5
- :H₂O
- :MFCD00011332
- :18.02 g/mol
- Pakning:Plastic bottle
- Pakn:125 mL