Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Vand, DEPC treated

Vand, DEPC treated

Plastic bottle, 1 L
Katalog # 786-109
Leverandør:   G-Biosciences
Vand, DEPC treated
Katalog # 786-109
Leverandør:  G-Biosciences
CAS nummer:  7732-18-5
Vand, DEPC treated
Vand, DEPC treated

Plastic bottle, 1 L

CAS nummer:  7732-18-5
Price
575,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Ultra pure DEPC-Treated water is suitable for use with sensitive RNA research. It is prepared by incubating with 0.1% DEPC (diethylpyrocarbonate), an RNase inhibitor, and then autoclaved to remove the DEPC, to prevent downstream interference, and sterilized.

Synonymer:  Hydrogen oxide
  • CAS nummer:7732-18-5
  • Formel:H₂O
  • MDL nummer:MFCD00011332
  • Molecular Weight:18.02 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.