Gel loading dyes, Glow™
Glow™ dye 6X, BromoBlue™, 1,5 mL
Katalog # 786-103
Leverandør: G-Biosciences
Gå til:
Product Details & Documents
These premixed and ready-to-use gel dyes are designed to minimise the use of ethidium bromide. Glow dyes provide intense DNA/RNA bands with little background or band distortion on any type of agarose or acrylamide gel.
- Dyes for DNA are Ficoll®-based
- Glow loading dyes already contain ethidium bromide
- Both RNA loading dyes are formamide-based
- Glow RNA for quick screenings of RNA preps
Specifically formulated for nucleic acid electrophoresis.
Specifikationer
Specifikationer
- Pakn:1,5 mL
- Beskrivelse:Glow™ dye 6X, BromoBlue™