Polysorbat 80 NF, Multi-Compendial, J.T.Baker®
Glass bottle, amber, narrow mouth, 1 L
: Avantor
- GMP manufactured product
- Vegetable based
Suitable for use in the manufacture of parenteral dosage forms.
: Sorbitanmono(cis-9-octadecenoat), ethoxyliert
- :9005-65-6
- :MFCD00082107
- :–21 °C
- :1,062 g/cm³
- :Management of Change (MOC) category = R
- :113 °C (closed cup)
- :Stue temperatur
- :500-019-9
Documents
sds_BAKR4117
SDS
spec_BAKR4117-02 spec_BAKR4117-03 spec_BAKR4117-04 spec_BAKR4117-04 spec_BAKR4117-06
Product specification
- Meets E.P. Chemical Specifications:≤2.0
- Meets B.P. Chemical Specifications:65 - 80
- Meets N.F. Requirements:Passes Test
- Meets J.P. Chemical Specifications:Passes Test
- GMP Manufactured Product:≤0.25%
- CAUTION: For Manufacturing, processing or repackaging:45 - 55
- Bulk Pharmaceutical Chemical:1.06 - 1.09
- NF - Acid Value:≤ 2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤ 0.25 %
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25°C:1.06 - 1.09
- NF - Viscosity at 25°C, cSt:300 - 500
- NF - Water (H₂O):≤ 3.0 %
- NF - Peroxide Value:≤ 10.0
- NF - Ethylene Oxide:≤ 1 ppm
- NF - Dioxane:≤ 10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- NF - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- NF - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- NF - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- NF - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- NF - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- EP - Acid Value:≤ 2.0
- EP - Total Ash:≤ 0.25 %
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤ 10.0
- EP - Ethylene Oxide:≤ 1 ppm
- EP - Dioxan:≤ 10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤ 3.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- Appearance:Passes Test
- JP - Acid Value:≤ 2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- JP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- JP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- JP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- JP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- JP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- JP - Dioxane:≤ 10 ppm
- JP - Ethylene Oxide:≤ 1 ppm
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤ 10.0
- JP - Residue on Ignition:≤ 0.25 %
- JP - Water (H₂O):≤ 3.0 %
- JP - Saponification Value:45 - 55
- Additional Tests - Color (Gardner):≤ 7
- Additional Tests - Odor (Faint):Passes Test
- Additional Tests - Peroxide Value, meqO₂/kg:≤ 2.0
- Additional Tests - Endotoxin Concentration (EU/mL):≤ 10
- Free Ethylene Oxide:≤ 1 ppm
- 1,4-Dioxane:≤ 5 ppm
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤ 100
- Microbiological - Escherichia Coli:Passes Test
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Microbiological - Total Yeast and Mold Count (cfu/g):≤ 50
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol :≤ 0.10 %
- Pakning:Glass bottle, amber, narrow mouth
- Pakn:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



