2-Propanol USP, J.T.Baker®
4 L
Leverandør: Avantor
2-Propanol USP, J.T.Baker®
Katalog # 9080-03
Leverandør: Avantor
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Kogende Pt:82 °C (1013 hPa)
- Smeltende Pt:–89 °C
- Tæthed:0,786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-661-7
- FN:1219
- ADR:3,II
- NÅ:01-2119457558-25
Specifikationer
Specifikationstestresultater
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.783 - 0.787
- USP - Refractive Index, n²ºD:1.376 - 1.378
- USP - Acidity:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Assay:≥99.0 %
- USP - Water (H₂O):≤0.5 %
- USP - Volatile Impurities - Ethyl ether:≤0.1 %
- USP - Volatile Impurities - Acetone:≤0.1 %
- USP - Volatile Impurities - Diisopropyl ether:≤0.1 %
- USP - Volatile Impurities - 1-Propanol:≤0.1 %
- USP - Volatile Impurities - 2-Butanol:≤0.1 %
- USP - Volatile Impurities - Total:≤1.0 %
- USP - Benzene:≤2 ppm
Specifikationer
- Pakn:4 L
- Cat. No.:9080-03
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



