2-Propanol USP, J.T.Baker®
Glass bottle, 500 mL
Leverandør: Avantor
2-Propanol USP, J.T.Baker®
Katalog # 9080-01
Leverandør: Avantor
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Kogende Pt:82 °C (1013 hPa)
- Smeltende Pt:–89 °C
- Tæthed:0,786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-661-7
- FN:1219
- ADR:3,II
- NÅ:01-2119457558-25
Documents
Certifikater
Søg efter partinummerP20_BAKR9080-03_SPEC_EN.pdf spec_BAKR9080-01 spec_BAKR9080-R
Product specification
Specifikationer
Specifikationstestresultater
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.783 - 0.787
- USP - Refractive Index, n²ºD:1.376 - 1.378
- USP - Acidity:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Assay:≥99.0 %
- USP - Water (H₂O):≤0.5 %
- USP - Volatile Impurities - Ethyl ether:≤0.1 %
- USP - Volatile Impurities - Acetone:≤0.1 %
- USP - Volatile Impurities - Diisopropyl ether:≤0.1 %
- USP - Volatile Impurities - 1-Propanol:≤0.1 %
- USP - Volatile Impurities - 2-Butanol:≤0.1 %
- USP - Volatile Impurities - Total:≤1.0 %
- USP - Benzene:≤2 ppm
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:9080-01
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



