Hand towels, SCOTT®
SCOTT®, rulle med enkelt ark, hvid, 304 m × 200 mm, 6 ruller/kasse
Catalog # 115-2233
Supplier: KIMBERLY CLARK
Jump to:
Product Details & Documents
Disposable hand towels made from AIRFLEX* (a voluminous, textile-like fabric).
- High absorption performance, and tear-proof even when wet
- Single, self-presenting towels for enhanced hygiene and less waste
- Extremely soft
Specifications
- Pakket:6 ruller/kasse
- Farve:hvid
- Beskrivelse:SCOTT®, rulle med enkelt ark
- L×B:304 m × 200 mm
- Seq:05
Accessories
Product Information
- Catalog #BeskrivelseB×D×HAvailabilityPrice
- 115-1779Dispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with window326×241×430 mmLog in for availability1 * 1 Items - 1.480,00 kr