Orders LinkContactUsLinkComponent
Glass stoppers with hexagonal grip
14/23, Brunt glas
Katalog # WITG0.303.014
Leverandør:   witeg Labortechnik
Glass stoppers with hexagonal grip
Katalog # WITG0.303.014
Leverandør:  witeg Labortechnik
Glass stoppers with hexagonal grip
Glass stoppers with hexagonal grip
14/23, Brunt glas
Price
43,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
  • Acc. to DIN 12252
Specifikationer
  • NS:
    14/23
  • Model:
    Brunt glas