Glass stoppers with hexagonal grip
14/23, Brunt glas
Katalog # WITG0.303.014
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
- Acc. to DIN 12252
Specifikationer
- NS:14/23
- Model:Brunt glas