Orders LinkContactUsLinkComponent
Dunke
20 L, UN approval: UN 3H1/Y/1.9, 245×295×385 mm, GL 51
Katalog # 216-1040
Leverandør:   KARTELL
Dunke
Katalog # 216-1040
Leverandør:  KARTELL
Dunke
Dunke
20 L, UN approval: UN 3H1/Y/1.9, 245×295×385 mm, GL 51
Price
1.220,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
HDPE, translucent with black screw cap.
  • With comfortable handle and secure seal cap following DIN recommendations
  • Conform to UN regulation on transportation of dangerous liquids
  • Suitable for foodstuff
Ideal for storing and transporting liquids.
Specifikationer
  • Gevind:
    GL 51
  • Kapacitet:
    20 L
  • Beskrivelse:
    UN approval: UN 3H1/Y/1.9
  • B×D×H:
    245×295×385 mm